Bicine ≥99%
Supplier: BeanTown Chemical
Synonyms:
2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine
214435-1KG
214435-250G
214435-50G
BT214435-1KGEA
443.19
USD
BT214435-1KG
BT214435-250G
BT214435-50G
Bicine ≥99%
Bicine
CAS: 150-25-4; EC No: 205-755-1; MDL No: MFCD00004295; RTECS: MB9700000
Powder; Molecular Formula: C6H13NO4; MW: 163.17
- RTECS: MB9700000
- Powder
Formula:
C₆H₁₃NO₄ |
MDL Number:
MFCD00004295 CAS Number: 150-25-4 EINECS: 205-755-1 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...