Benzophenone hydrazone ≥98%
Supplier: BeanTown Chemical
214385-1KG
214385-250G
214385-50G
BT214385-1KGEA
251
USD
BT214385-1KG
BT214385-250G
BT214385-50G
Benzophenone hydrazone ≥98%
Benzophenone hydrazone
CAS: 5350-57-2; EC No: 226-321-8; MDL No: MFCD00007624; RTECS: PC4954487
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C13H12N2 ; MW: 196.25
Melting Point: 95-98°; Boiling Point: 225-230°/55 mmHg
- RTECS: PC4954487
- Crystalline
Formula:
C₁₃H₁₂N₂ Boiling Pt: 225…230 °C/55 mmHg Melting Pt: 95…98 °C |
MDL Number:
MFCD00007624 CAS Number: 5350-57-2 EINECS: 226-321-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...