Sulfur ≥99.9995%, pieces

Supplier: BeanTown Chemical

214375-100G 214375-25G 214375-500G
BT214375-100GEA 216 USD
BT214375-100G BT214375-25G BT214375-500G
Sulfur ≥99.9995%, pieces
Sulphur

CAS: 7704-34-9; EC No: 231-722-6; MDL No: MFCD00085316; RTECS: WS4250000 UN No: UN1350; Haz Class: 4.1; Packing Group: III Solid; Molecular Formula: S; MW: 32.07 Melting Point: 117-120°; Boiling Point: 444.7° Density (g/mL): 2.07


  • RTECS: WS4250000
  • Solid

Formula: S
Boiling Pt: 444.7 °C
Melting Pt: 117…120 °C
Density: 2.07 g/mL
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.9995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR