Phenyl vinyl sulfone ≥99%

Supplier: BeanTown Chemical

Synonyms: (Vinylsulfonyl)benzene, Vinyl phenyl sulfone, Vinyl phenyl sulphone, (Vinylsulphonyl)benzene

214370-1G 214370-25G 214370-5G
BT214370-1GEA 31 USD
BT214370-1G BT214370-25G BT214370-5G
Phenyl vinyl sulfone ≥99%
Phenyl vinyl sulphone

CAS: 5535-48-8; EC No: 226-890-2; MDL No: MFCD00007554 Crystalline; Linear Formula: C6H5SO2CH=CH2; Molecular Formula: C8H8O2S; MW: 168.22 Melting Point: 67-71°


  • Crystalline

Formula: C₈H₈O₂S
MW: 168.22 g/mol
Melting Pt: 67…71 °C
MDL Number: MFCD00007554
CAS Number: 5535-48-8
EINECS: 226-890-2

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR