Rhodium(III) chloride hydrate
Supplier: BeanTown Chemical
214270-1G
214270-250MG
214270-5G
BT214270-1GEA
436
USD
BT214270-1G
BT214270-250MG
BT214270-5G
Rhodium(III) chloride hydrate
Rhodium (III) chloride hydrate
CAS: 20765-98-4; MDL No: MFCD00149839
Crystalline; Linear Formula: RhCl3·xH2O; MW: 209.26 (anhydrous)
Melting Point: 100° (decomposes)
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
Cl₃Rh·H₂O Melting Pt: 100 °C (dec.) |
MDL Number:
MFCD00149839 CAS Number: 20765-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...