N-Z-L-Aspartic anhydride ≥95%
Supplier: BeanTown Chemical
214255-25G
BT214255-25GEA
133
USD
BT214255-25G
N-Z-L-Aspartic anhydride ≥95%
N-Z-L-Aspartic anhydride
CAS: 4515-23-5; EC No: 224-838-3; MDL No: MFCD00136820
Powder; Molecular Formula: C12H11NO5; MW: 249.22
Melting Point: 123-124°
Optical Rotation: [α]20/D -25°, c = 1 in methanol
- Optical rotation: [α]20/D -25°, c = 1 in methanol
- Powder
Formula:
C₁₂H₁₁NO₅ Melting Pt: 123…124 °C |
CAS Number:
4515-23-5 EINECS: 224-838-3 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...