(Diethylamino)sulfur trifluoride ≥95%
Supplier: BeanTown Chemical
Synonyms:
DAST
214105-100G
214105-25G
214105-5G
BT214105-100GEA
682
USD
BT214105-100G
BT214105-25G
BT214105-5G
(Diethylamino)sulfur trifluoride ≥95%
(Diethylamino)sulphur trifluoride
CAS: 38078-09-0; EC No: 253-771-2; MDL No: MFCD00000363
UN No: UN2920; Haz Class: 8 (3); Packing Group: I
Liquid; Linear Formula: (CH3CH2)2NSF3; Molecular Formula: C4H10F3NS; MW: 161.19
Boiling Point: 30-32°/3 mmHg; Flash point: 23°C (73°F)
Density (g/mL): 1.22
Moisture Sensitive
- Moisture Sensitive
- Liquid
Formula:
C₄H₁₀F₃NS Boiling Pt: 30…32 °C/3 mmHg Density: 1.22 g/mL Flash Pt: 23 °C (73 °F) |
MDL Number:
MFCD00000363 CAS Number: 38078-09-0 EINECS: 253-771-2 UN: 2920 ADR: 8,I |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...