Aluminium nitrate hydrate ≥99.999%

Supplier: BeanTown Chemical

214055-100G 214055-25G 214055-5G
BT214055-100GEA 236.27 USD
BT214055-100G BT214055-25G BT214055-5G
Aluminium nitrate hydrate ≥99.999%
Aluminium nitrate hydrate

CAS: 25838-59-9; EC No: 236-751-8; MDL No: MFCD00003419 UN No: UN1438; Haz Class: 5.1; Packing Group: III Crystalline; Linear Formula: Al(NO3)3·xH2O ; MW: 212.99 (anhydrous) Melting Point: 73° Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: AlN₃O₉·H₂O
MW: 231.01 g/mol
Melting Pt: 73 °C
MDL Number: MFCD00003419
CAS Number: 25838-59-9
EINECS: 236-751-8
UN: 1438
ADR: 5.1 III

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR