Calcium sulfate, Drierite™ 10-20 mesh for drying, without indicator

Supplier: BeanTown Chemical

Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum, Calcium sulfate (anhydrous) anhydrite

214005-1LB 214005-5LB
BT214005-1LBEA 41.55 USD
BT214005-1LB BT214005-5LB
Calcium sulfate, Drierite™ 10-20 mesh for drying, without indicator
Calcium sulphate

CAS: 7778-18-9; EC No: 231-900-3; MDL No: MFCD00010912 Granular Moisture Sensitive


  • Moisture Sensitive
  • Granular

Formula: CaO₄S
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR