α-D-Lactose monohydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
α-Lactose monohydrate
214000-1KG
214000-5KG
BT214000-1KGEA
50.56
USD
BT214000-1KG
BT214000-5KG
α-D-Lactose monohydrate ≥99%
α-D-Lactose monohydrate
CAS: 5989-81-1; MDL No: MFCD00150747
Solid; Molecular Formula: C12H22O11·H2O; MW: 360.31
Melting Point: 219°
- Solid
Formula:
C₁₂H₂₂O₁₁·H₂O Melting Pt: 219 °C |
MDL Number:
MFCD00150747 CAS Number: 5989-81-1 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...