1-Propanol ≥99.0% for HPLC

Supplier: BeanTown Chemical

Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

213970-1L 213970-4L 213970-4X1L
BT213970-1LEA 70.32 USD
BT213970-1L BT213970-4L BT213970-4X1L
1-Propanol ≥99.0% for HPLC
1-Propanol

CAS: 71-23-8; EC No: 200-746-9; MDL No: MFCD00002941; RTECS: UH8225000 UN No: UN1274; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3CH2CH2OH; Molecular Formula: C3H8; MW: 60.10 Melting Point: -127°; Boiling Point: 97°; Flash point: 22°C (72°F) Density (g/mL): 0.804; Refractive Index: 1.384


  • RTECS: UH8225000
  • Refractive index: 1.384
  • Liquid

Formula: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Boiling Pt: 97
Melting Pt: -127
Density: 0.804 g/mL
Flash Pt: 22 °C (72 °F)
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3 II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR