Strontium fluoride ≥99%

Supplier: BeanTown Chemical

213965-1KG 213965-250G 213965-5KG
BT213965-1KGEA 125 USD
BT213965-1KG BT213965-250G BT213965-5KG
Strontium fluoride ≥99%
Strontium fluoride

CAS: 7783-48-4; EC No: 232-000-3; MDL No: MFCD00011251; RTECS: WK8925000 Powder; Linear Formula: SrF2; MW: 125.62 Melting Point: 1473°; Boiling Point: 2489° Density (g/mL): 4.24; Refractive Index: 1.442


  • RTECS: WK8925000
  • Refractive index: 1.442
  • Powder

Formula: F₂Sr
Boiling Pt: 2489 °C
Melting Pt: 1473 °C
Density: 4.24 g/mL
MDL Number: MFCD00011251
CAS Number: 7783-48-4
EINECS: 232-000-3
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR