Tetrakis(triphenylphosphine)palladium(0) ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4
213825-10G
213825-2G
213825-500MG
BT213825-10GEA
272.36
USD
BT213825-10G
BT213825-2G
BT213825-500MG
Tetrakis(triphenylphosphine)palladium(0) ≥99.5%
Tetrakis(triphenylphosphine)palladium (0)
CAS: 14221-01-3; EC No: 238-086-9; MDL No: MFCD00010012
Crystalline; Linear Formula: Pd[P(C6H5)3]4; Molecular Formula: C72C60P4Pd; MW: 1155.57
Melting Point: 100-105°
Air Sensitive, Light Sensitive, Moisture Sensitive
- Air Sensitive, Light Sensitive, Moisture Sensitive
- Crystalline
Formula:
C₇₂H₆₀P₄Pd Melting Pt: 100…105 °C |
MDL Number:
MFCD00010012 CAS Number: 14221-01-3 EINECS: 238-086-9 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...