3-Picolyl chloride hydrochloride ≥97%

Supplier: BeanTown Chemical

Synonyms: 3-(Chloromethyl)-pyridinium chloride, 3-(Chloromethyl)pyridine hydrochloride

213785-100G 213785-25G 213785-5G
BT213785-100GEA 206 USD
BT213785-100G BT213785-25G BT213785-5G
3-Picolyl chloride hydrochloride ≥97%
3-Picolyl chloride hydrochloride

CAS: 6959-48-4; EC No: 230-150-4; MDL No: MFCD00012818; RTECS: US7000000 UN No: UN3261 ; Haz Class: 8; Packing Group: III Solid; Molecular Formula: C6H6ClN·HCl; MW: 164.03 Melting Point: 137-143° Hygroscopic


  • RTECS: US7000000
  • Hygroscopic
  • Solid

Formula: C₆H₇Cl₂N
MW: 164.03 g/mol
Melting Pt: 137…143 °C
MDL Number: MFCD00012818
CAS Number: 6959-48-4
EINECS: 230-150-4
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR