2,3,5-Trifluoropyridine ≥98%
Supplier: BeanTown Chemical
213705-1G
213705-25G
213705-5G
BT213705-1GEA
39
USD
BT213705-1G
BT213705-25G
BT213705-5G
2,3,5-Trifluoropyridine ≥98%
2,3,5-Trifluoropyridine
CAS: 76469-41-5; EC No: 000-000-0; MDL No: MFCD03001162
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C5H2F3N; MW: 133.07
Flash point: 30°C (86°F)
Density (g/mL): 1.400
- Liquid
Formula:
C₅H₂F₃N Density: 1.400 g/mL Flash Pt: 30 °C (86 °F) |
MDL Number:
MFCD03001162 CAS Number: 76469-41-5 UN: 1993 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...