Sodium carbonate, anhydrous ≥99.997%

Supplier: BeanTown Chemical

Synonyms: Soda, Carbonic acid sodium salt, Calcined soda, Disodium carbonate, Carbonic acid disodium salt, Na-X, Bisodium carbonate, Carbonic acid, disodium salt, Soda ash

213675-100G 213675-25G 213675-5G
BT213675-100GEA 473.93 USD
BT213675-100G BT213675-25G BT213675-5G
Sodium carbonate, anhydrous ≥99.997%
Sodium carbonate

CAS: 497-19-8; EC No: 207-838-8; MDL No: MFCD00003494; RTECS: VZ4050000 Powder; Linear Formula: Na2CO3; MW: 105.99 Melting Point: 851° Density (g/mL): 2.53; Refractive Index: 1.535 Hygroscopic


  • RTECS: VZ4050000
  • Refractive index: 1.535
  • Hygroscopic
  • Powder

Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854
Density: 2.53 g/mL
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.997%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR