Silica gel, granules 3-8 mesh desiccant

Supplier: BeanTown Chemical

213560-1KG 213560-250G 213560-5KG
BT213560-1KGEA 61.77 USD
BT213560-1KG BT213560-250G BT213560-5KG
Silica gel, granules 3-8 mesh desiccant
Silica gel

CAS: 63231-67-4; EC No: 231-545-4; MDL No: MFCD00011232 Solid Melting Point: 1600°; Boiling Point: 2230° Hygroscopic


  • Hygroscopic
  • Solid

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: >1600 °C
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR