Thiourea ≥99%

Supplier: BeanTown Chemical

Synonyms: Sulfourea, Thiocarbamide, Sulphourea

213505-100G 213505-2.5KG 213505-500G
BT213505-100GEA 36 USD
BT213505-100G BT213505-2.5KG BT213505-500G
Thiourea ≥99%
Thiourea

CAS: 62-56-6; EC No: 200-543-5; MDL No: MFCD00008067; RTECS: YU2800000 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline; Linear Formula: NH2CSNH2; Molecular Formula: CH4N2S; MW: 76.12 Melting Point: 170-176° Density (g/mL): 1.405


  • RTECS: YU2800000
  • Crystalline

Formula: NH₂CSNH₂
MW: 76.12 g/mol
Melting Pt: 170…176 °C
Density: 1.405 g/mL
MDL Number: MFCD00008067
CAS Number: 62-56-6
EINECS: 200-543-5
UN: 3077
ADR: 9 III
Merck Index: 13,09443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&vid=e9ae5dc49e92d68f893244a5cd4e0245f289ed09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR