(4-Methylbenzyl)triphenylphosphonium chloride ≥98%
Supplier: BeanTown Chemical
213450-10G
213450-250G
213450-50G
BT213450-10GEA
41
USD
BT213450-10G
BT213450-250G
BT213450-50G
(4-Methylbenzyl)triphenylphosphonium chloride ≥98%
(4-Methylbenzyl)triphenylphosphonium chloride
CAS: 1530-37-6; MDL No: MFCD00031581
Solid; Molecular Formula: C26H24ClP; MW: 402.91
- Solid
Formula:
C₂₆H₂₄ClP MW: 402.91 g/mol |
MDL Number:
MFCD00031581 CAS Number: 1530-37-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...