Silicon carbide ≥99% (trace metals basis), whiskers
Supplier: BeanTown Chemical
213385-10G
213385-250G
213385-50G
BT213385-10GEA
62.97
USD
BT213385-10G
BT213385-250G
BT213385-50G
Silicon carbide ≥99% (trace metals basis), whiskers
Silicon carbide
CAS: 409-21-2; EC No: 206-991-8; MDL No: MFCD00049531
Needles, 1.5 micron dia, ~18 micron length; Linear Formula: SiC; MW: 40.10
Melting Point: 2700°
Density (g/mL): 3.22
- Needles, 1.5 micron dia, ~18 micron length
Formula:
SiC Melting Pt: 2700 °C Density: 3.22 g/mL |
MDL Number:
MFCD00049531 CAS Number: 409-21-2 EINECS: 206-991-8 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...