Silicon carbide ≥99% (trace metals basis), whiskers

Supplier: BeanTown Chemical

213385-10G 213385-250G 213385-50G
BT213385-10GEA 62.97 USD
BT213385-10G BT213385-250G BT213385-50G
Silicon carbide ≥99% (trace metals basis), whiskers
Silicon carbide

CAS: 409-21-2; EC No: 206-991-8; MDL No: MFCD00049531 Needles, 1.5 micron dia, ~18 micron length; Linear Formula: SiC; MW: 40.10 Melting Point: 2700° Density (g/mL): 3.22


  • Needles, 1.5 micron dia, ~18 micron length

Formula: SiC
Melting Pt: 2700 °C
Density: 3.22 g/mL
MDL Number: MFCD00049531
CAS Number: 409-21-2
EINECS: 206-991-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd532b391b9c28edcb8adac5fe5ed958ccaa487&vid=d39ce1cfd6c1a5ca44c7d11b656758cbfaed190d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR