Arsenic (V) oxide ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Arsenic oxide, Arsenic pentoxide, di-Arsenic pentaoxide
213285-100G
213285-25G
BT213285-100GEA
424
USD
BT213285-100G
BT213285-25G
Arsenic (V) oxide ≥99.9%
Arsenic (V) oxide
CAS: 1303-28-2; EC No: 215-116-9; MDL No: MFCD00003434; RTECS: CG2275000
UN No: UN1559; Haz Class: 6.1; Packing Group: II
-100 Mesh Powder; Molecular Formula: As2O5; MW: 229.79
Melting Point: 315° (decomposes)
Hygroscopic
- RTECS: CG2275000
- Hygroscopic
- -100 Mesh Powder
Formula:
As₂O₅ Melting Pt: 315 °C (dec.) |
MDL Number:
MFCD00003434 CAS Number: 1303-28-2 EINECS: 215-116-9 UN: 1559 ADR: 6.1,II |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...