3-(Trimethylsilyl)-2-propyn-1-ol ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-(Trimethylsilyl)propargyl alcohol

213235-1G 213235-25G 213235-5G
BT213235-1GEA 32 USD
BT213235-1G BT213235-25G BT213235-5G
3-(Trimethylsilyl)-2-propyn-1-ol ≥98%
3-(Trimethylsilyl)-2-propyn-1-ol

CAS: 5272-36-6; EC No: 226-094-5; MDL No: MFCD00002913 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C6H12OSi; MW: 128.24 Boiling Point: 76°/11 mmHg; Flash point: 67°C (153°F) Density (g/mL): 0.865; Refractive Index: 1.451 Moisture Sensitive


  • Refractive index: 1.451
  • Moisture Sensitive
  • Liquid

Formula: C₆H₁₂OSi
MW: 128.25 g/mol
Boiling Pt: 76 °C/11 mmHg
Density: 0.865 g/mL
Flash Pt: 67 °C (153 °F)
MDL Number: MFCD00002913
CAS Number: 5272-36-6
EINECS: 226-094-5
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR