Pentaethylene glycol ≥98%

Supplier: BeanTown Chemical

Synonyms: 3,6,9,12-Tetraoxatetradecane-1,14-diol

213205-10G 213205-250G 213205-50G
BT213205-10GEA 127.85 USD
BT213205-10G BT213205-250G BT213205-50G
Pentaethylene glycol ≥98%
Pentaethylene glycol

CAS: 4792-15-8; EC No: 225-341-4; MDL No: MFCD00002878; RTECS: RZ2670000 Liquid; Linear Formula: H(OCH2CH2)5OH; Molecular Formula: C10H22O6; MW: 238.28 Boiling Point: 338-340°; Flash point: 110°C (230°F) Density (g/mL): 1.126; Refractive Index: 1.462 Hygroscopic


  • RTECS: RZ2670000
  • Refractive index: 1.462
  • Hygroscopic
  • Liquid

Formula: C₁₀H₂₂O₆
Boiling Pt: 338…340 °C
Density: 1.126 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00002878
CAS Number: 4792-15-8
EINECS: 225-341-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR