2,5-Dibromobenzoic acid ≥98%
Supplier: BeanTown Chemical
213160-25G
213160-5G
BT213160-25GEA
242
USD
BT213160-25G
BT213160-5G
2,5-Dibromobenzoic acid ≥98%
2,5-Dibromobenzoic acid
CAS: 610-71-9; EC No: 210-234-7; MDL No: MFCD00016494; RTECS: DG6290000
Solid; Molecular Formula: C7H4Br2O2 ; MW: 279.91
Melting Point: 156-159°
- RTECS: DG6290000
- Solid
Formula:
C₇H₄Br₂O₂ Melting Pt: 156…159 °C |
MDL Number:
MFCD00016494 CAS Number: 610-71-9 EINECS: 210-234-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...