1,4-Dioxaspiro[4.5]decan-8-one ≥97%

Supplier: BeanTown Chemical

Synonyms: 1,4-Cyclohexanedione monoethylene acetal, 1,4-Cyclohexanedione monoethylene ketal

213065-100G 213065-25G 213065-5G
BT213065-100GEA 448 USD
BT213065-100G BT213065-25G BT213065-5G
1,4-Dioxaspiro[4.5]decan-8-one ≥97%
1,4-Dioxaspiro[4.5]decan-8-one

CAS: 4746-97-8; MDL No: MFCD00010214 Crystalline; Molecular Formula: C8H12O3; MW: 156.18 Melting Point: 70-73° Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: C₈H₁₂O₃
MW: 156.18 g/mol
Melting Pt: 70…73 °C
MDL Number: MFCD00010214
CAS Number: 4746-97-8

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR