9-Fluorenylmethanol ≥99%

Supplier: BeanTown Chemical

Synonyms: (Fluoren-9-yl)methanol, 9-Fluorenmethanol, 9-Fluorenemethanol

213005-100G 213005-25G 213005-5G
BT213005-100GEA 593 USD
BT213005-100G BT213005-25G BT213005-5G
9-Fluorenylmethanol ≥99%
9-Fluorenylmethanol

CAS: 24324-17-2; EC No: 246-167-5; MDL No: MFCD00001139 Crystalline; Molecular Formula: C14H12O ; MW: 196.24 Melting Point: 105-107°


  • Crystalline

Formula: C₁₄H₁₂O
Melting Pt: 105…107 °C
MDL Number: MFCD00001139
CAS Number: 24324-17-2
EINECS: 246-167-5

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR