6-Thioguanine ≥98%

Supplier: BeanTown Chemical

212935-1G 212935-5G
BT212935-1GEA 82.43 USD
BT212935-1G BT212935-5G
6-Thioguanine ≥98%
6-Thioguanine

CAS: 154-42-7; EC No: 205-827-2; MDL No: MFCD00233553; RTECS: UP0740000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C5H5N5S; MW: 167.19 Melting Point: 300°


  • RTECS: UP0740000
  • Powder

Formula: C₅H₅N₅S
Melting Pt: >300 °C
MDL Number: MFCD00233553
CAS Number: 154-42-7
EINECS: 205-827-2
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR