Quercetin dihydrate ≥97%
Supplier: BeanTown Chemical
Synonyms:
2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate
212870-100G
212870-25G
212870-500G
BT212870-100GEA
99
USD
BT212870-100G
BT212870-25G
BT212870-500G
Quercetin dihydrate ≥97%
Quercetin dihydrate
CAS: 6151-25-3; EC No: 204-187-1; MDL No: MFCD00149487; RTECS: LK8950000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C15H10O7·2H2O; MW: 338.28
Melting Point: 310-315°
- RTECS: LK8950000
- Powder
Formula:
C₁₅H₁₄O₉ Melting Pt: 310…315 °C |
MDL Number:
MFCD00149487 CAS Number: 6151-25-3 EINECS: 204-187-1 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...