Manganese ≥99.95% (trace metals basis), powder -325 mesh
Supplier: BeanTown Chemical
212840-100G
212840-25G
BT212840-100GEA
190.95
USD
BT212840-100G
BT212840-25G
Manganese ≥99.95% (trace metals basis), powder -325 mesh
Manganese
CAS: 7439-96-5; EC No: 231-105-1; MDL No: MFCD00011111; RTECS: OO9625000
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: Mn; MW: 54.94
Melting Point: 1244°; Boiling Point: 1962°
Density (g/mL): 7.3
- RTECS: OO9625000
- Powder
Formula:
Mn Boiling Pt: 1962 °C Melting Pt: 1244 °C Density: 7.3 g/mL |
MDL Number:
MFCD00011111 CAS Number: 7439-96-5 EINECS: 231-105-1 UN: 3089 ADR: 4.3,III |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...