(4-Fluorophenyl)acetic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-(4-Fluorophenyl)acetic acid

212750-100G 212750-25G 212750-5G
BT212750-100GEA 220 USD
BT212750-100G BT212750-25G BT212750-5G
(4-Fluorophenyl)acetic acid ≥98%
(4-Fluorophenyl)acetic acid

CAS: 405-50-5; EC No: 206-972-4; MDL No: MFCD00004343 Crystalline; Molecular Formula: C8H7FO2; MW: 154.14 Melting Point: 81-83°


  • Crystalline

Formula: (C₆H₄F)CH₂COOH
Melting Pt: 81…83 °C
MDL Number: MFCD00004343
CAS Number: 405-50-5
EINECS: 206-972-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR