Bromotrimethylsilane ≥97% stabilised

Supplier: BeanTown Chemical

Synonyms: Trimethylbromosilane, Trimethylsilyl bromide

212740-10G 212740-250G 212740-50G
BT212740-10GEA 53.77 USD
BT212740-10G BT212740-250G BT212740-50G
Bromotrimethylsilane ≥97% stabilised
Bromotrimethylsilane

CAS: 2857-97-8; EC No: 220-672-0; MDL No: MFCD00000048 UN No: UN2924; Haz Class: 3(8); Packing Group: II Liquid; Linear Formula: (CH3)3SiBr; MW: 153.09 Boiling Point: 79°; Flash point: 25°C (77°F) Density (g/mL): 1.16; Refractive Index: 1.424 Moisture Sensitive


  • Refractive index: 1.424
  • Moisture Sensitive
  • Liquid

Formula: C₃H₉BrSi
MW: 153.09 g/mol
Boiling Pt: 79
Melting Pt: -43
Density: 1.16 g/mL
Flash Pt: 25 °C (77 °F)
MDL Number: MFCD00000048
CAS Number: 2857-97-8
EINECS: 220-672-0
UN: 2924
ADR: 3 II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR