4-(Boc-amino)-2-chloropyridine ≥95%

Supplier: BeanTown Chemical

Synonyms: 2-Chloro-4-(tert-butoxycarbonyl)pyridine, tert-Butyl (2-chloropyridin-4-yl)carbamate, 4-Amino-2-chloropyridine, 4-BOC protected, tert-Butyl 2-chloropyridine-4-carbamate

212730-1G 212730-5G
BT212730-1GEA 205 USD
BT212730-1G BT212730-5G
4-(Boc-amino)-2-chloropyridine ≥95%
4-(Boc-amino)-2-chloropyridine

CAS: 234108-73-7; MDL No: MFCD02179232 Crystalline/Powder; Molecular Formula: C10H13ClN2O2 ; MW: 228.68


  • Crystalline/Powder

Formula: C₁₀H₁₃ClN₂O₂
MDL Number: MFCD02179232
CAS Number: 234108-73-7

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR