Ethyl hydrogen malonate ≥90%, Technical Grade

Supplier: BeanTown Chemical

212700-100G 212700-25G 212700-5G
BT212700-100GEA 317 USD
BT212700-100G BT212700-25G BT212700-5G
Ethyl hydrogen malonate ≥90%, Technical Grade
Ethyl hydrogen malonate

CAS: 1071-46-1; EC No: 213-992-7; MDL No: MFCD00020490 Liquid; Linear Formula: HO2CCH2CO2CH2CH3; Molecular Formula: C5H8O4; MW: 132.12 Boiling Point: 106°/3 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.119; Refractive Index: 1.435


  • Refractive index: 1.435
  • Liquid

Formula: C₅H₈O₄
Boiling Pt: 106 °C/3 mmHg
Density: 1.119 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00020490
CAS Number: 1071-46-1
EINECS: 213-992-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR