Platinum : rhodium (90:10) (w/w), wire 0.5 mm dia. for ISA type R standard grade thermocouple
Supplier: BeanTown Chemical
212685-100CM
212685-25CM
BT212685-100CMEA
852
USD
BT212685-100CM
BT212685-25CM
Platinum : rhodium (90:10) (w/w), wire 0.5 mm dia. for ISA type R standard grade thermocouple
Platinum : rhodium
MDL No: MFCD00198186
Solid; Formula: Pt:Rh; 90:10 wt%
- Solid