Salicylic acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
2-Hydroxybenzoic acid, o-Hydroxybenzoic acid
212665-250G
212665-500G
BT212665-250GEA
18.74
USD
BT212665-250G
BT212665-500G
Salicylic acid ≥99%
Salicylic acid
CAS: 69-72-7; EC No: 200-712-3; MDL No: MFCD00002439; RTECS: VO0525000
Crystalline; Linear Formula: HOC6H4COOH; Molecular Formula: C7H6O3; MW: 138.12
Melting Point: 158-161°; Boiling Point: 211°/20 mmHg; Flash point: 157°C (315°F)
Density (g/mL): 1.443
Light Sensitive
- RTECS: VO0525000
- Light Sensitive
- Crystalline
Formula:
(HO)C₆H₄CO₂H Boiling Pt: 211 °C/20 mmHg Melting Pt: 158…161 °C Density: 1.443 g/mL Flash Pt: 157 °C (315 °F) |
MDL Number:
MFCD00002439 CAS Number: 69-72-7 EINECS: 200-712-3 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...