5-Hydroxy-1-indanone ≥95%
Supplier: BeanTown Chemical
212660-5G
BT212660-5GEA
63
USD
BT212660-5G
5-Hydroxy-1-indanone ≥95%
5-Hydroxy-1-indanone
CAS: 3470-49-3; MDL No: MFCD00857527
Solid; Molecular Formula: C9H8O2; MW: 148.16
Melting Point: 175° (decomposes)
- Solid
Formula:
C9H8O2 Melting Pt: 175 °C (dec.) |
MDL Number:
MFCD00857527 CAS Number: 3470-49-3 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...