(3-Chlorophenyl)acetic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-Chlorophenylacetic acid

212515-100G 212515-25G 212515-5G
BT212515-100GEA 327 USD
BT212515-100G BT212515-25G BT212515-5G
(3-Chlorophenyl)acetic acid ≥98%
(3-Chlorophenyl)acetic acid

CAS:1878-65-5 ; EC No: 217-520-0; MDL No: MFCD00004332 Powder; Molecular Formula: C8H7ClO2; MW: 170.59 Melting Point: 76-79°


  • Powder

Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Boiling Pt: 292
Melting Pt: 76…79 °C
MDL Number: MFCD00004332
CAS Number: 1878-65-5
EINECS: 217-520-0
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR