Neopentyl bromide ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,2-Dimethyl-1-propyl bromide, 1-Bromo-2,2-dimethylpropane

212450-25G 212450-5G
BT212450-25GEA 200 USD
BT212450-25G BT212450-5G
Neopentyl bromide ≥98%
Neopentyl bromide

CAS: 630-17-1; EC No: 211-132-5; MDL No: MFCD00000209 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)3CCH2Br; MW: 151.05 Boiling Point: 104-106°; Flash point: 6°C (42°F) Density (g/mL): 1.199; Refractive Index: 1.436


  • Refractive index: 1.436
  • Liquid

Formula: C₅H₁₁Br
Boiling Pt: 104…106 °C
Density: 1.199 g/mL
Flash Pt: 6 °C (42 °F)
MDL Number: MFCD00000209
CAS Number: 630-17-1
EINECS: 211-132-5
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR