Sodium iodate ≥99%

Supplier: BeanTown Chemical

212330-100G 212330-500G
BT212330-100GEA 50.38 USD
BT212330-100G BT212330-500G
Sodium iodate ≥99%
Sodium iodate

CAS: 7681-55-2; EC No: 231-672-5; MDL No: MFCD00003533 UN No: UN1479; Haz Class: 5.1; Packing Group: II Powder; Linear Formula: NaIO3; MW: 197.89 Density (g/mL): 4.28 Air Sensitive, Light Sensitive, Moisture Sensitive


  • Air Sensitive, Light Sensitive, Moisture Sensitive
  • Powder

Formula: INaO₃
Density: 4.28 g/mL
MDL Number: MFCD00003533
CAS Number: 7681-55-2
EINECS: 231-672-5
UN: 1479
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=806173453dcec1d2f791367d269d662d3459149a&vid=16d5efbf45a7cc1cd6399272211e2455e47a2e4b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR