Formylhydrazine ≥98%
Supplier: BeanTown Chemical
Synonyms:
Formic acid hydrazide, Formohydrazide, Formic hydrazide
212190-100G
212190-25G
BT212190-100GEA
155
USD
BT212190-100G
BT212190-25G
Formylhydrazine ≥98%
Formylhydrazine
CAS: 624-84-0; EC No: 210-867-9; MDL No: MFCD00007589; RTECS: LQ8615000
Flakes; Linear Formula: HCONHNH2; Molecular Formula: C4HN2O; MW: 60.06
Melting Point: 52-57°; Flash point: 112°C (234°F)
Hygroscopic
- RTECS: LQ8615000
- Hygroscopic
- Flakes
Formula:
CH₄N₂O MW: 60.06 Boiling Pt: 90 Melting Pt: 150 Flash Pt: 112 °C (234 °F) |
MDL Number:
MFCD00007589 CAS Number: 624-84-0 EINECS: 210-867-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...