Potassium hexacyanoferrate(II) trihydrate 98.5-102.0% ACS

Supplier: BeanTown Chemical

212135-100G 212135-2KG 212135-500G
BT212135-100GEA 51.81 USD
BT212135-100G BT212135-2KG BT212135-500G
Potassium hexacyanoferrate(II) trihydrate 98.5-102.0% ACS
Potassium hexacyanoferrate (II) trihydrate

CAS: 14459-95-1; EC No: 237-722-2; MDL No: MFCD00167023 Crystalline; Linear Formula: K4Fe(CN)6·3H2O; MW: 422.39 Melting Point: 69-71° Density (g/mL): 1.85


  • Crystalline

Formula: K₄Fe(CN)₆·3H₂O
Melting Pt: 69…71 °C
Density: 1.85 g/mL
MDL Number: MFCD00167023
CAS Number: 14459-95-1
EINECS: 237-722-2

Order Now

Specification Test Results

Assay 98.5-102.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR