Sodium methoxide ≥98%
Supplier: BeanTown Chemical
Synonyms:
Sodium methylate, Sodium methanolate
212100-100G
212100-2KG
212100-500G
BT212100-100GEA
37.49
USD
BT212100-100G
BT212100-2KG
BT212100-500G
Sodium methoxide ≥98%
Sodium methoxide
CAS: 124-41-4; EC No: 204-699-5; MDL No: MFCD00012179; RTECS: PC3570000
UN No: UN1431; Haz Class: 4.2 (8); Packing Group: II
Powder; Linear Formula: CH3ONa; MW: 54.02
Melting Point:
- RTECS: PC3570000
- Moisture Sensitive
- Powder
Formula:
CH₃NaO MW: 54.02 g/mol Melting Pt: 127 Density: 0.97 g/mL Flash Pt: 33 °C (91 °F) |
MDL Number:
MFCD00012179 CAS Number: 124-41-4 EINECS: 204-699-5 UN: 1431 ADR: 4.2 II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...