Ethyl-4-iodobenzoate ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethyl 4-iodobenzoate, 4-Iodobenzoic acid ethyl ester

212085-10G 212085-250G 212085-50G
BT212085-10GEA 50 USD
BT212085-10G BT212085-250G BT212085-50G
Ethyl-4-iodobenzoate ≥98%
Ethyl 4-iodobenzoate

CAS: 51934-41-9; EC No: 429-550-3; MDL No: MFCD00017344 Liquid; Molecular Formula: C9H9IO2; MW: 276.08 Flash point: 110°C (230°F) Density (g/mL): 1.641; Refractive Index: 1.588 Light Sensitive


  • Refractive index: 1.588
  • Light Sensitive
  • Liquid

Formula: C₉H₉IO₂
Density: 1.641 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00017344
CAS Number: 51934-41-9
EINECS: 429-550-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR