Titanium ≥99.99% (trace metals basis), foil, Thickness 0.25 mm

Supplier: BeanTown Chemical

212080-100X100 212080-25X25MM 212080-50X50MM
BT212080-100X1EA 716 USD
BT212080-100X1 BT212080-25X25 BT212080-50X50
Titanium ≥99.99% (trace metals basis), foil, Thickness 0.25 mm
Titanium

CAS: 7440-32-6; EC No: 231-142-3; MDL No: MFCD00011264; RTECS: XR1700000 Solid; Molecular Formula: Ti; MW: 47.87 Melting Point: 1660°; Boiling Point: 3287° Density (g/mL): 4.5


  • RTECS: XR1700000
  • Solid

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.5 g/mL
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR