Sodium tetrachloroaurate dihydrate ≥99.99%

Supplier: BeanTown Chemical

Synonyms: Sodium tetrachloroaurate(III) dihydrate

212045-1G 212045-5G
BT212045-1GEA 173.76 USD
BT212045-1G BT212045-5G
Sodium tetrachloroaurate dihydrate ≥99.99%
Sodium tetrachloroaurate dihydrate

CAS: 13874-02-7; EC No: 239-241-3; MDL No: MFCD00149162 UN No: UN3260; Haz Class: 8; Packing Group: III Solid; Linear Formula: NaAuCl4·2H2O; MW: 397.80 Melting Point: 100° (decomposes) Density (g/mL): 0.8


  • Solid

Formula: NaAuCl₄·2H₂O
MW: 397.8 g/mol
Melting Pt: 100 °C (dec.)
Density: 0.8 g/mL
MDL Number: MFCD00149162
CAS Number: 13874-02-7
EINECS: 239-241-3
UN: 3260
ADR: 8 III

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR