Chromium(III) fluoride ≥99.98%

Supplier: BeanTown Chemical

Synonyms: Chromium trifluoride

211830-1G 211830-5G
BT211830-1GEA 82 USD
BT211830-1G BT211830-5G
Chromium(III) fluoride ≥99.98%
Chromium (III) fluoride

CAS: 7788-97-8; EC No: 232-137-9; MDL No: MFCD00016045 UN No: UN1756; Haz Class: 8; Packing Group: II Powder; Molecular Formula: CrF3; MW: 108.99 Density (g/mL): 3.8 Hygroscopic


  • Hygroscopic
  • Powder

Formula: CrF₃
Density: 3.8 g/mL
MDL Number: MFCD00016045
CAS Number: 7788-97-8
EINECS: 232-137-9
UN: 1756
ADR: 8,II

Order Now

Specification Test Results

Assay 99.98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR