di-Sodium L(+)-tartrate dihydrate 99.0-101.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Sodium L-(+)-tartrate dihydrate, L(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Sodium tartrate dibasic dihydrate, Disodium L-(+)-Tartrate Dihydrate, (R,R)-(+)-Disodium tartrate dihydrate, L(+)-Tartaric acid, disodium salt, dihydrate, Sodium L-tartrate dibasic dihydrate, L(+)-Sodium tartrate dibasic dihydrate, Sodium Tartrate, Dihydrate, (+)-Disodium L-tartrate dihydrate, (+)-Disodium-L-tartrat dihydrate
CAS: 6106-24-7; EC No: 212-773-3; MDL No: MFCD00150035; RTECS: WW8150000
Granular; Molecular Formula: C4H4Na2O6·2H2O; MW: 230.08
Density (g/mL): 1.820; Optical Rotation: [α]20/D +26°, c = 1 in water
- RTECS: WW8150000
- Optical rotation: [α]20/D +26°, c = 1 in water
- Granular
Formula:
NaOOC(CHOH)₂COONa·2H₂O Density: 1.820 g/mL |
MDL Number:
MFCD00150035 CAS Number: 6106-24-7 EINECS: 212-773-3 |
|
Specification Test Results
Assay | 99.0-101.0% |
Loss on drying (150°C) | 15.61-15.71% |
pH of 5% solution at 25°C | 7.0-9.0 |
Insoluble matter | ≤0.005% |
Chloride (Cl) | ≤0.0005% |
Phosphate (PO4) | ≤0.0005% |
Sulfate (SO4) | ≤0.005% |
Ammonium (NH4) | ≤0.003% |
Calcium (Ca) | ≤0.01% |
Heavy metals (as Pb) | ≤0.0005% |
Iron (Fe) | ≤0.001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...