(Ethyl)triphenylphosphonium bromide ≥98%
Supplier: BeanTown Chemical
Synonyms:
Ethyltriphenylphosphonium bromide
211730-100G
211730-25G
211730-500G
BT211730-100GEA
89
USD
BT211730-100G
BT211730-25G
BT211730-500G
(Ethyl)triphenylphosphonium bromide ≥98%
(Ethyl)triphenylphosphonium bromide
CAS: 1530-32-1; EC No: 216-223-3; MDL No: MFCD00011838
UN No: UN3077; Haz Class: 9; Packing Group: III
Solid; Molecular Formula: C20H20BrP ; MW: 371.26
Melting Point: 203-205°
Hygroscopic
- Hygroscopic
- Solid
Formula:
C₂₀H₂₀BrP MW: 371.26 g/mol Melting Pt: 203…205 °C |
MDL Number:
MFCD00011838 CAS Number: 1530-32-1 EINECS: 216-223-3 UN: 3077 ADR: 6.1 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...