4-Vinylphenylboronic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: Dihydroxy(4-vinylphenyl)borane, (4-Vinylphenyl)metaboric acid, 4-Vinylbenzeneboronic acid

211650-1G 211650-25G 211650-5G
BT211650-1GEA 35 USD
BT211650-1G BT211650-25G BT211650-5G
4-Vinylphenylboronic acid ≥97%
4-Vinylphenylboronic acid

CAS: 2156-04-9; MDL No: MFCD00239441 Solid; Molecular Formula: C8H9BO2 ; MW: 147.97 Melting Point: 190-193°


  • Solid

Formula: C₈H₉BO₂
MW: 147.97 g/mol
Melting Pt: 190…193 °C
MDL Number: MFCD00239441
CAS Number: 2156-04-9

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR